Orders LinkContactUsLinkComponent
VWR®, インクジェットプリンタスライド
VWR®, インクジェットプリンタスライド
Supplier:   VWR Collection
VWR®, インクジェットプリンタスライド
Supplier:  VWR Collection
Certificates
VWR®, インクジェットプリンタスライド
VWR®, インクジェットプリンタスライド
4 Options Available Below
Product Details & Documents
Microscopy slides are designed with colour-frosted ends for H&E stain on frozen sections in histopathology examination.
  • Shelf life of 60 months
  • Pre-cleaned, without mark, print or contamination risks
  • Marking with conventional labelling system, pencil, marker pens and inkjet printers
Specially produced for all inkjet printers.
包装: 50 pieces per box. 50 boxes per case.
注意: Fragile items should be stored in dry and proper laboratory temperature conditions.
Product Family Options
Product Information
  • 説明
    Colour
    Material
    厚さ
    長さ×幅
    Availability
    Price
  • Inkjet printer coloured-frosted slides on 20 mm
    White
    Glass
    1,0 (-0,05/+0,1) mm
    76×25 mm
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
  • Inkjet printer coloured-frosted slides on 20 mm
    Blue
    Glass
    1,0 (-0,05/+0,1) mm
    76×25 mm
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
  • Inkjet printer coloured-frosted slides on 20 mm
    Green
    Glass
    1,0 (-0,05/+0,1) mm
    76×25 mm
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items
  • Inkjet printer coloured-frosted slides on 20 mm
    Yellow
    Glass
    1,0 (-0,05/+0,1) mm
    76×25 mm
    Log in for availability
    Log in for availability
    1 * 2.500 items
    1 * 2.500 items