Orders LinkContactUsLinkComponent
VWR®, 保護フードとフェースカバー
VWR®, 保護フードとフェースカバー
VWR®, 保護フードとフェースカバー
Supplier:  VWR Collection
Certificates
VWR®, 保護フードとフェースカバー
VWR®, 保護フードとフェースカバー
2 Options Available Below
Product Details & Documents
These poypropylene head and face covers are designed to protect users from potential environmental contaminations.
  • Bulk packaging
  • White colour
Weight: 20 g
Delivery information: Supplied in a case of 500, garments are bulk packed with polybag and case liner.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8202
  • 114-9456
  • Garment application:
  • Laboratory
  • Laboratory
Specifications
Product Family Options
Product Information
  • 説明
    Colour
    衣服のサイズ
    Availability
    Price
  • Hood and face cover
    White
    L
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8202
    Garment application
    Laboratory
  • Hood and face cover
    White
    XL
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
    More
    Specifications
    Cat. No.
    114-9456
    Garment application
    Laboratory