Orders LinkContactUsLinkComponent
VWR®, 細胞培養用プレート, 3D scaffold
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 細胞培養用プレート, 3D scaffold
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 細胞培養用プレート, 3D scaffold
3 Options Available Below
Product Details & Documents
VWR® 3D scaffold cell culture plates, made of polystyrene (GPPS) with polystyrene polymer scaffold, are able to simulate the three-dimensional structure of cells in animals and the human body to maximum effect, providing an ideal environment for cell interaction, maximising culture area, and improving yield.
  • Highly consistent scaffold (mean fibre Ø of 500 and 260 μm pore spacing) with 3D channels providing high connectivity, facilitating nutrient transmission, consistent metabolic activity and the accuracy of culture results
  • Cytokine and growth factor resistant surface assists cell secretion collection, saving time and reducing the need for additional separating steps
  • Open pores with high connectivity facilitate nutrient absorption and metabolism
  • Larger surface area than regular cell culture dishes for greater yields and maximum material efficiency
  • Strictly integrity tested
  • Sterilised by electron beam irradiation
  • Free from DNase and RNase, non pyrogenic
Applications include studies of the cell-cell interaction mechanism, cell immunotherapy, stem cell therapy, drug screening and the production of cell-based drugs.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2970
  • 734-2971
  • 734-2972
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 説明
    Culture area
    Packed
    Availability
    Price
  • Standard 6-well plate, 3× surface treated scaffold inserts (33,5×1,6 mm)
    143 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 8 items
    1 * 8 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2970
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Standard 12-well plate, 6× surface treated scaffold inserts (21,0×1,6 mm)
    113 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 8 items
    1 * 8 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2971
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Standard 24-well plate, 12× surface treated scaffold inserts (15,0×1,6 mm)
    115 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 8 items
    1 * 8 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2972
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
Complementary Products
Product Information
  • Catalog #
    Packed
    Growth area
    説明
    直径
    Availability
    Price
  • 734-2967
    Individually wrapped
    43 cm²
    Standard dish, with 1× surface treated scaffold insert (32,0×1,6 mm)
    35 mm
    Log in for availability
    1 * 40 items - $0.00