Orders LinkContactUsLinkComponent
VWR®, 培養ディッシュにおける細胞スライド
VWR®, 培養ディッシュにおける細胞スライド
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 培養ディッシュにおける細胞スライド
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 培養ディッシュにおける細胞スライド
VWR®, 培養ディッシュにおける細胞スライド
4 Options Available Below
Product Details & Documents
VWR® cell slides are made from medical grade, high transparency PET, which has high strength and is unbreakable. Only the surface of the cell slide has a hydrophilic treatment so adherent cells grow easily, whilst the surrounding culture dish surface is hydrophobic, making it difficult for adherent cells to grow.
  • Cell slide transparency and light transmittance are good so cells can be clearly observed under a light or fluorescence microscope
  • The use of multiple cell slides in one dish makes it possible to develop multi-factor, multi-index, multi-level in vitro studies under the same condition
  • Choice of Ø 8 or Ø 10 mm cell slides, with 12, 18, 32 or 45 cell slides per culture dish
  • One cell culture can prepare multiple cell slides for different studies and objectives, increasing work efficiency
  • Tab on the cell slide tilts at an angle, which is convenient for operators to directly clamp, and is engraved numerically, facilitating identification
  • Free from DNase and RNase, non pyrogenic
The integrated cell slides are fixed in the culture dish. If further testing is necessary after cell culture, the cell slides can be taken out one by one and placed in a culture plate or other container with the necessary specifications.
Delivery information: Supplied individually and packed in 'peel to open' plastic blister packaging. Each box is matched with individually packed sterile metal tweezers. Both cell slides and tweezers are electron beam irradiated and should be used immediately after opening.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2973
  • 734-2974
  • 734-2975
  • 734-2976
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 説明
    直径
    Growth area
    Packed
    Availability
    Price
  • Culture dish, 60 mm, with 18 cell slides, cell growth area 0,50 cm² per cell slide
    8 mm
    9,00 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 48 items
    1 * 48 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2973
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Culture dish, 60 mm, with 12 cell slides, cell growth area 0,79 cm² per cell slide
    10 mm
    9,42 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 48 items
    1 * 48 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2974
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Culture dish, 100 mm, with 45 cell slides, cell growth area 0,50 cm² per cell slide
    8 mm
    22,50 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2975
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Culture dish, 100 mm, with 32 cell slides, cell growth area 0,79 cm² per cell slide
    10 mm
    25,12 cm²
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2976
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M