Orders LinkContactUsLinkComponent
VWR®, ガラスビーズ
Supplier:   VWR Collection
VWR®, ガラスビーズ
Supplier:  VWR Collection
Certificates
VWR®, ガラスビーズ
VWR®, ガラスビーズ
10 Options Available Below
Product Details & Documents
Soda-lime glass, solid.
  • Not suitable for heat applications
  • Available in sizes from 1 to 10 mm
Can be used for many different applications in the laboratory. In microbiology laboratories they are used for the production of single cell suspensions or for plating. In chemical laboratories they are used as filling material for distillation columns. They can also be used to clean round bottom vessels.
Product Family Options
Product Information
  • 説明
    直径
    Availability
    Price
  • Soda-lime glass
    1,0 - 1,3 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 2,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 3,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 4,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 5,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 6,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 7,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 8,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 9,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • 10,0 mm
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g