Orders LinkContactUsLinkComponent
VWR® CTP, クリーンルーム用テープ
VWR® CTP, クリーンルーム用テープ
Supplier:   VWR Collection
VWR® CTP, クリーンルーム用テープ
Supplier:  VWR Collection
Certificates
VWR® CTP, クリーンルーム用テープ
VWR® CTP, クリーンルーム用テープ
2 Options Available Below
Product Details & Documents
These tapes are made of polyethylene, with a synthetic rubber adhesive.
  • Easily bonds different media
  • Serrated edge for easy tear
  • High tack adhesive
Length: 32,9 m
Product Family Options
Product Information
  • 説明
    Colour
    Availability
    Price
  • Cleanroom construction tape, CTP
    White
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 36 Roll
    1 * 36 Roll
  • Cleanroom construction tape, CTP
    White
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll