Orders LinkContactUsLinkComponent
VWR®, ビーカー、低形状、PP
VWR®, ビーカー、低形状、PP
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, ビーカー、低形状、PP
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, ビーカー、低形状、PP
9 Options Available Below
Product Details & Documents
PP, translucent.
  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.
Specifications for Product Family
Specifications
  • Catalog No:
  • 213-0511
  • 213-0489
  • 213-0490
  • 213-0491
  • 213-0492
  • 213-0493
  • 213-0494
  • 213-0495
  • 213-0496
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Specifications
Product Family Options
Product Information
  • 容量
    Availability
    Price
  • 25 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0511
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0489
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 100 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0490
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0491
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0492
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0493
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 2000 ml
    1 * 1 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0494
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 3000 ml
    1 * 1 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0495
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 5000 ml
    1 * 1 items
    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    213-0496
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging