Avantor SterilEnz®-II/AT: pre-gasketed sanitary fittings for single-use systems
Catalog # 734-3209
Supplier: PAW BIOSCIENCE
Specifications
- 説明:SterilEnz®-II/AT standard sanitary fitting with gasket, 3/8" HB × 1" TC fitting size
- Material:PP
- Seq:03
- Pk:10
Specifications
About this item
SterilEnz®-II/AT connectors come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face. They are designed for use in irradiated disposable systems.
- Standard sanitary clamp fitting, per ASME BPE Specifications
- Class VI Compliant materials; certificate included
- Won't leak or blow-off like stepped barbs can
- Mono-barb design
- Zero mold parting lines on all critical sealing surfaces
- Superior tooling
- Gamma-stable, 25 to 40 kGY
Certifications: USP <88>: Class VI Biological reactivity tests, in vivo
USP <87>: Cytotoxicity, MEM Elution, in vitro
USP <661>: Physicochemical Test for plastics
USP <1207>: Sterile Product Packaging, hose barb integrity evaluation, via microbial immersion
ASTM F756-00 : Rabbit blood cell hemolysis
SterilEnz®-II/AT are certified ADCF (animal derived component free).
All SterilEnz®-II/AT products are assembled inside a certified ISO 5 (Class 100) environment.
Protected by U.S. Patent 7,950,700, other patents pending. SterilEnz® is a registered trademark of PAW BioScience Products, LLC.
USP <87>: Cytotoxicity, MEM Elution, in vitro
USP <661>: Physicochemical Test for plastics
USP <1207>: Sterile Product Packaging, hose barb integrity evaluation, via microbial immersion
ASTM F756-00 : Rabbit blood cell hemolysis
SterilEnz®-II/AT are certified ADCF (animal derived component free).
All SterilEnz®-II/AT products are assembled inside a certified ISO 5 (Class 100) environment.
Protected by U.S. Patent 7,950,700, other patents pending. SterilEnz® is a registered trademark of PAW BioScience Products, LLC.