Orders LinkContactUsLinkComponent
VWR®, ビーカー、使い捨て
VWR®, ビーカー、使い捨て
VWR®, ビーカー、使い捨て
Supplier:  VWR Collection
Certificates
VWR®, ビーカー、使い捨て
VWR®, ビーカー、使い捨て
9 Options Available Below
Product Details & Documents
PP, translucent.
  • Embossed graduations
  • Ribs for easy stacking
  • Autoclavable to 121 °C
  • With spout
High quality economical beaker for everyday work. Ideal for general laboratory use.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • 容量
    高さ
    外径
    目盛り範囲
    Availability
    Price
  • 15 ml
    38 mm
    37 mm
    5 - 15 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 30 ml
    44 mm
    46 mm
    5 - 30 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 50 ml
    52 mm
    52 mm
    5 - 50 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 100 ml
    62 mm
    64 mm
    5 - 100 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 150 ml
    79 mm
    70 mm
    5 - 150 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 250 ml
    90 mm
    80 mm
    10 - 250 ml
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • 400 ml
    111 mm
    96 mm
    25 - 400 ml
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • 400 ml
    111 mm
    96 mm
    25 - 400 ml
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - $75.27
    $75.27
    1 * 50 items
       $75.27
  • 600 ml
    124 mm
    107 mm
    25 - 600 ml
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items