Orders LinkContactUsLinkComponent
VWR®, 顕微鏡用共焦点ディッシュ
VWR®, 顕微鏡用共焦点ディッシュ
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 顕微鏡用共焦点ディッシュ
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 顕微鏡用共焦点ディッシュ
2 Options Available Below
Product Details & Documents
Confocal dishes are suitable for fluorescent, confocal and phase-contrast microscopy experiments. These dishes combine the convenience of standard 35 mm, PS cell culture dishes with the optical benefits of glass. Dishes are available with Ø 20 mm glass centre, with or without a tissue culture-treated surface.
  • Made from homogenous borosilicate glass with low bubble and inclusion content
  • Transparent, medical grade glue, achromatic
  • Dish surface is smooth and free from striations to maximise usable area for growth
  • Rim on upper side of the lid mates with the dish brim for easy and secure stacking
  • Vented lids for effective gas exchange
  • Suitable for all living cell examinations
  • Sterilised by electron beam irradiation and non-pyrogenic
Glass thickness: 0,16 to 0,19 mm.
 : ISO 13485:2003 and ISO 9001:2008 certified.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2904
  • 734-2906
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 説明
    直径
    Availability
    Price
  • Glass bottom dish, Ø20 mm centre, TC-treated
    35 mm
    Log in for availability
    Log in for availability
    1 * 40 items
    1 * 40 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2904
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Glass bottom dish, Ø20 mm centre, non treated
    35 mm
    Log in for availability
    Log in for availability
    1 * 40 items
    1 * 40 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2906
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact