Orders LinkContactUsLinkComponent
VWR®, dNTPs
VWR®, dNTPs
Supplier:  VWR Chemicals
Certificates
VWR®, dNTPs
4 Options Available Below
Product Details & Documents
Ready to use molecular biology grade dNTP mixes and dNTP sets. The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. dNTP solutions are also available in sets of four individual dNTPs, each 100 mm. Both are convenient for use in DNA polymerisation reactions, DNA labelling and sequencing processes.
  • Available as pre-mixed 10 or 25 mM solutions, or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7,3 to 7,5
Product Family Options
Product Information
  • 説明
    Pk
    Availability
    Price
  • dNTP mix, 10 mM of each dA, dC, dG, and dT, 2×500 μl
    1.000 µl
    Log in for availability
    Log in for availability
    1 * 1.000 µl
    1 * 1.000 µl - $229.93
    $229.93
    1 * 1.000 µl
       $229.93
  • dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 4×250 μl
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - $524.02
    $524.02
    1 * 1 SET
       $524.02
  • dNTP mix, 25 mM of each dA, dC, dG, and dT, 2×1 ml
    2.000 µl
    Log in for availability
    Log in for availability
    1 * 2.000 µl
    1 * 2.000 µl - $854.99
    $854.99
    1 * 2.000 µl
       $854.99
  • dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 16×250 μl
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - $1,893.74
    $1,893.74
    1 * 1 SET
       $1,893.74