Orders LinkContactUsLinkComponent
試験ストリップ、塩化物、Dosatest
試験ストリップ、塩化物、Dosatest
Chloride test 0 - 3000 mg/l, 0 · 500 · 1000 · 1500 · 2000 · ≥3000 mg/l, 100
Catalog # 85441.601
Supplier:   VWR Chemicals
試験ストリップ、塩化物、Dosatest
Catalog # 85441.601
Supplier:  VWR Chemicals
CAS Number:  
試験ストリップ、塩化物、Dosatest
試験ストリップ、塩化物、Dosatest
Chloride test 0 - 3000 mg/l, 0 · 500 · 1000 · 1500 · 2000 · ≥3000 mg/l, 100
CAS Number:  
Price
$72.02
Log in for availability
1 * 100 items
Product Details & Documents
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Ideal to control your disinfection. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
包装: Container with 100 test strips 6 x 95 mm.
Documents
Specifications
Specifications
  • Pk:
    100
  • 説明:
    Chloride test 0 - 3000 mg/l
  • Range:
    0 · 500 · 1000 · 1500 · 2000 · ≥3000 mg/l
Specifications