Orders LinkContactUsLinkComponent
試験ストリップ、亜硝酸塩、Dosatest
試験ストリップ、亜硝酸塩、Dosatest
Nitrite test strips 0 - 80 mg/l, 0 · 1 · 5 · 10 · 20 · 40 · 80, 100
Catalog # 85440.601
Supplier:   VWR Chemicals
試験ストリップ、亜硝酸塩、Dosatest
Catalog # 85440.601
Supplier:  VWR Chemicals
CAS Number:  
試験ストリップ、亜硝酸塩、Dosatest
試験ストリップ、亜硝酸塩、Dosatest
Nitrite test strips 0 - 80 mg/l, 0 · 1 · 5 · 10 · 20 · 40 · 80, 100
CAS Number:  
Price
$81.82
Log in for availability
1 * 100 items
Product Details & Documents
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
 : Container with 100 test strips 6 x 95 mm.
Documents
Specifications
Specifications
  • Pk:
    100
  • 説明:
    Nitrite test strips 0 - 80 mg/l
  • Range:
    0 · 1 · 5 · 10 · 20 · 40 · 80
Specifications