Orders LinkContactUsLinkComponent
試験ストリップ、過酸化物、Dosatest
試験ストリップ、過酸化物、Dosatest
試験ストリップ、過酸化物、Dosatest
Supplier:  VWR Chemicals
Certificates
試験ストリップ、過酸化物、Dosatest
試験ストリップ、過酸化物、Dosatest
3 Options Available Below
Product Details & Documents
A ready to use kit that fits into your lab coat pocket. Intended for water-based samples
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.
Product Family Options
Product Information
  • 説明
    Range
    Pk
    Availability
    Price
  • Peroxide test strips 0 - 25 mg/l
    0 · 0,5 · 2 · 5 · 10 · 25
    100
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $58.80
    $58.80
    1 * 100 items
       $58.80
  • Peroxide test strips 0 - 100 mg/l
    0 · 1 · 3 · 10 · 30 · 100
    100
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $57.39
    $57.39
    1 * 100 items
       $57.39
  • Peroxide test strips 0 - 1000 mg/l
    0 · 50 · 150 · 300 · 500 · 800 · 1000
    100
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $82.23
    $82.23
    1 * 100 items
       $82.23