pHインジケーターペーパー、1~14、Rota
Catalog # 85402.600
Supplier: VWR Chemicals
Specifications
- pH gradation:1 · 2 · 3 · 5 · 6 · 7 · 8 · 9 · 10 · 12 · 14
- pH range:1 - 14
- Pk:5
Specifications
About this item
Indicator paper for testing across the complete pH range.
- Easy to use
- Produced from high quality paper to give good colour matching of paper to printed calibration scale
Delivery: Supplied in plastic reels that ensure protection and long term stability. Reel of 5 m length, width: 7 mm.