LiChrosorb®、RP-8, HPLCカラム
Supplier: HICHROM
About this item
These columns with high purity base deactivated porous silica are ideal for the separation of basic molecules.
- Irregular porous silica
- High efficiency and complimentary phase selectivity
8 Options Available Below
Product Details & Documents
These columns with high purity base deactivated porous silica are ideal for the separation of basic molecules.
- Irregular porous silica
- High efficiency and complimentary phase selectivity
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- LIRP8-5-125A
- LIRP8-5-150A
- LIRP8-5-250A
- LIRP8-10-125AF
- LIRP8-10-250AF
- LIRP8-10-200A
- LIRP8-10-250A
- LIRP8-5-10C5
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9,5%
- 9.5%
- Endcapped:
- No|No
- No|No
- No|No
- No|No
- No|No
- No|No
- No|No
- No|No
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
Product Information
- カラム形式Phase細孔サイズ粒子サイズ内径長さAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-5-125AEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-5-150AEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-5-250AEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-10-125AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-10-250AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-10-200AEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-10-250AEnvironmentally PreferableLow Manufacturing ImpactCarbon load9,5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard cartridges
Product Information
- カラム形式Phase細孔サイズ粒子サイズ内径長さAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.LIRP8-5-10C5Environmentally PreferableLow Manufacturing ImpactCarbon load9.5%EndcappedNo|NoSurface area300 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact