Orders LinkContactUsLinkComponent
VWR®, 乳棒、細胞ストレイナー用
VWR®, 乳棒、細胞ストレイナー用
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 乳棒、細胞ストレイナー用
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 乳棒、細胞ストレイナー用
2 Options Available Below
Product Details & Documents
Wear-proof purple polypropylene pestles which are designed to fit into cell strainers perfectly.
  • Grooved handle for ease of use
  • Sterilised by electron beam irradiation
  • Non-pyrogenic, DNase/RNase free
Packaging: Each package has lot number printed on it to facilitate quality tracking. Supplied as individually packaged in easy to tear paper and plastic for easy opening (100/case). Bulk packaging has 25/bag and 50/case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2819
  • 734-2820
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 説明
    For
    Colour
    Availability
    Price
  • Pestle, sterile, individually packaged
    Cell strainers
    Purple
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2819
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Pestles, sterile, bulk packaging
    Cell strainers
    Purple
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2820
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M