Orders LinkContactUsLinkComponent
VWR®, 濾過装置
VWR®, 濾過装置
VWR®, 濾過装置
Supplier:  VWR Collection
Certificates
VWR®, 濾過装置
2 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • Very good temperature resistance
  • Available with 1000 or 2000 ml flask
  • Funnel with 300 or 500 ml capacity
All-glass filtration apparatus, with funnel, fritted base, cap, 47 mm Ø spring clamp and ground joint flask. This unit is recommended for general filtration. Applications include filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction discs. Support base has a sealed-in, coarse porosity, fritted disc.
Product Family Options
Product Information
  • 説明
    容量
    Material
    Availability
    Price
  • With 1000 ml flask
    300 ml
    Borosilicate glass 3.3
    1 * 1 items
  • With 2000 ml flask
    500 ml
    Borosilicate glass 3.3
    1 * 1 items
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0667
    Spare gasket, PTFE, 47 mm, for filtration apparatus
    Log in for availability
    1 * 5 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0269
    Spring clamp, aluminium, 47 mm for filtration apparatus
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0270
    Spare glass base and cap for filtration apparatus, 47 mm
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0271
    Spare ground joint flask for filtration apparatus, 1000 ml capacity
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0267
    Spare glass funnel 300 ml for filtration apparatus, borosilicate 3.3 glass
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0268
    Spare glass funnel 500 ml for filtration apparatus, borosilicate 3.3 glass
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 511-0272
    Spare ground joint flask for filtration apparatus, 2000 ml capacity