VWR® Talon® Support Stands, Heavy Duty
Catalog # VWRU60100-105
Supplier: VWR Collection
Specifications
- 説明:Support stand, with base (432×330 mm) and rod (16×457 mm)
- Seq:01
- Pk:1
Specifications
About this item
Multi-purpose support was originally designed to permit cylindrical vessels up to 305mm (12") in diameter to sit close to the upright rod.
- Versatile
- Heavy-duty design
- Multi-purpose application
- Supplied with stainless steel rod
- Finished in black crackle lacquer
Support is ideal for mounting stirring motors or for heavy equipment. Each of the four corners rests on a rubber cushion. Supplied with stainless steel rod 457mm (18") high, 16mm (5/8") diameter.