Sulphuric acid 0.5 mol/l (1 N) in aqueous solution USP Test Solution (TS) 34
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Storage Temperature:Ambient
EINECS:0
UN:3264
ADR:8,III