Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
クロージャー
Bottle Closures
VWR®, Screw Caps for Sampling Bottles
VWR®, Screw Caps for Sampling Bottles
VWR®, Screw Caps for Sampling Bottles
331-0322
:
VWR Collection
VWR®, Screw Caps for Sampling Bottles
331-0322
:
VWR Collection
For:
Sampling bottles
Thread:
28 mm
Colour:
White
説明:
Screw caps, tamper evident
Material:
PP
Pk:
5.000