VWR®, プレパラート、種子植物
Prepared slides with phanerogams
Catalog # 720-2926
Supplier: VWR Collection
Jump to:
Product Details & Documents
Prepared slides of seed bearing plants (phanerogams), including.
- Pine tree leaf (c.s.)
- Pine tree, 1 year old stem (c.s.)
- Pine tree, 10 year old stem (c.s.)
- Pine tree, spherical male fruit (l.s.)
- Pine tree, spherical female fruit (l.s.)
- Pine tree, pollen (w.b.)
- Buttercup root (c.s.)
- Corn (mais) root (c.s.)
- Mitosis, onion tips (s.)
- Geranium stem (c.s.)
- Marchantia antheridiophore (l.s.)
- Marchantia archegoniophore (l.s.)
- Sunflower, young stem (c.s.)
- Tilia (linden tree), 1 year old stem (c.s.)
- Tilia (linden tree), 3 year old stem (c.s.)
- Lily leaf (c.s.)
- Fig tree, leaf with foveolate stomata (c.s.)
- Three types of leaves (c.s.)
- Lily pollen (w.b.)
- Lily anther (c.s.)
- Lily ovary (c.s.)
- Meiosis of lily anther at different phases (c.s.)
- Capsella, young embryo (s.)
- Capsella, medium embryo (s.)
- Capsella, old embryo (s.)
(w.b.) = whole body; (s.) section; (c.s.) cross section; (l.s.) longitudinal section
Documents
720-2926_EN
Datasheet
Specifications
- 説明:Prepared slides with phanerogams