VWR®, 安全メガネ、ブダペスト
Anti-UV (2-1,2), anti-scratch, anti-fog, Clear PC
111-1847
Wrap-around protective eyewear combining fashion, fit and function. One-piece PC lens and a lightweight, frameless design ensures a comfortable custom fit all day long.
- Adjustable temple length enables wearers to fit the eyewear to their facial profile
- Pantoscopic adjustment allows lens angle modification
- Integrated side shield
- Soft nose bridge for extra comfort
: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
- Type:Anti-UV (2-1,2), anti-scratch, anti-fog
- Lenses:Clear PC
Accessories
- 説明長さ×幅Packed
- 説明
- 説明
- 説明
- 説明
- 説明
- 説明