VWR®, 安全メガネ、プラハ
Anti-UV, anti-scratch, anti-fog, Blue, Clear PC
Catalog # 111-1846
Supplier: VWR Collection
Jump to:
Product Details & Documents
These economical, compact cover glasses (OTG) fit over most prescription eyewear.
- Pantoscopic adjustment allows lens angle modification
- Adjustable temple length
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
Documents
Specifications
- Type:Anti-UV, anti-scratch, anti-fog
- Colour:Blue
- Lenses:Clear PC
Accessories
Product Information
- Catalog #説明AvailabilityPrice
Product Information
- Catalog #説明長さ×幅PackedAvailabilityPrice