VWR®, 安全メガネ、ロンドン
Anti-UV (2-1,2), anti-scratch, anti-fog, Light blue, Clear PC
Catalog # 111-1845
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stylish protective eyewear combining fashion, fit and function.
- Moulded-in nose bridge
- Soft, non slip rubber temple tips
- Ideal for small facial contours
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
Documents
111-1845
Images 360
TDS Safety Spectacle London
Datasheet
VWR® Products for Electrophoresis VWR® Safety Eyewear
Product Literature
Specifications
- Type:Anti-UV (2-1,2), anti-scratch, anti-fog
- Colour:Light blue
- Lenses:Clear PC
- Cat. No.:111-1845
Accessories
Product Information
- Catalog #説明長さ×幅PackedAvailabilityPrice