VWR® 耐薬品性オーバーオール
Supplier: VWR Collection
Certificates
About this item
These robust yet lightweight overalls are made from an optimised non woven PE fabric. Its softer feel and open structure still offers adequate Type 5 and 6 barrier protection and remains permeable to both air and moisture vapour.
- Two-piece hood, zip closure with flap
- Elasticated wrists, ankles, face and waist (stitched-in)
- Anti-static treatment (EN 1149-1) on outside
- Stitched external seams in yellow for visual identification and differentiation
- Provides protection against radioactive contamination (EN 1073-2)
Colour: White
Weight: <180 g
Certifications: EN ISO 13982-1:2004 + A1:2010. EN 13034:2005 + A1:2009. EN 1149-5:2008. EN 1073-2:2002. Type 5/6, Cat. III.
Packaging: Individually packed, 25 garments plastic wrapped.
Specifications
- Catalog No:
- 113-8379
- 113-8381
- 113-8380
- 113-8384
- 113-8383
- 113-8382
- Anti-static:
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial