VWR®, 安全メガネ、バルセロナ
Anti-UV (2-1,2), anti-scratch, anti-fog, Grey/blue, Clear PC
Catalog # 111-1848
Supplier: VWR Collection
Jump to:
Product Details & Documents
Modern and stylish design glasses with sturdy polycarbonate frame.
- Nose pad made from soft material for extra comfort and to avoid slippage
- Soft temple dual-injected rubber to fit different head profiles
- Pantoscopic adjustment
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
Documents
Specifications
- Type:Anti-UV (2-1,2), anti-scratch, anti-fog
- Colour:Grey/blue
- Lenses:Clear PC
Accessories
Product Information
- Catalog #説明AvailabilityPrice
Product Information
- Catalog #説明AvailabilityPrice
Product Information
- Catalog #説明AvailabilityPrice
Product Information
- Catalog #説明AvailabilityPrice
Product Information
- Catalog #説明長さ×幅PackedAvailabilityPrice