Orders LinkContactUsLinkComponent
VWR®, 滴下瓶、
VWR®, 滴下瓶、
VWR®, 滴下瓶、
VWR®, 滴下瓶、
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Product Family Options
Dropping bottles
  • 説明
    容量
  • Bottle only
    5 ml
    1 * 228 items
    1 * 228 items
  • Bottle only
    10 ml
    1 * 198 items
    1 * 198 items
  • Bottle only
    15 ml
    1 * 156 items
    1 * 156 items
  • Bottle only
    20 ml
    1 * 143 items
    1 * 143 items
  • Bottle only
    30 ml
    1 * 110 items
    1 * 110 items
  • Bottle only
    50 ml
    1 * 144 items
    1 * 144 items
Pipette kit with blue bulb
  • 説明
    容量
  • Pipette kit for 5 ml dropping bottles
    -
    1 * 100 items
    1 * 100 items
  • Pipette kit for 10 and 15 ml dropping bottles
    -
    1 * 100 items
    1 * 100 items
  • Pipette kit for 20 ml dropping bottles
    -
    1 * 100 items
    1 * 100 items
  • Pipette kit for 30 ml dropping bottles
    -
    1 * 100 items
    1 * 100 items
  • Pipette kit for 50 ml dropping bottles
    -
    1 * 100 items
    1 * 100 items
  • Pipette kit for 100 ml dropping bottles