EDTA disodium salt dihydrate 0.1 M, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5632-02
Supplier: Avantor
CAS Number:
Specifications
- パックタイプ:PE bottle, narrow mouth
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.098 - 0.102
- Molarity (Mol/L):0.098 - 0.102
Specification Test Results
About this item
:
:372.24 g/mol
Density:1,02 g/cm³
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037