Order Entry
Japan
ContactUsLinkComponent
 
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate 98.0-102.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1670-04
Supplier:  Avantor
CAS Number:  
undefined
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate 98.0-102.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1670-04
Supplier:  Avantor
CAS Number:  

Specifications

  • パックタイプ:
    Plastic bottle for solids
  • Pk:
    125 g
  • Seq:
    3445

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 102.0 %
  • Assay (CoCl₂6H₂O) (by EDTA titrn):
    98.0 - 102.0 %
  • Insoluble Matter:
    ≤ 0.01 %
  • Nitrate (NO₃):
    ≤ 0.01 %
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Calcium (Ca):
    ≤ 0.005 %
  • Copper (Cu):
    ≤ 0.002 %
  • Iron (Fe):
    ≤ 0.005 %
  • Magnesium (Mg):
    ≤ 0.005 %
  • Nickel (Ni):
    ≤ 0.1 %
  • Potassium (K):
    ≤ 0.01 %
  • Sodium (Na):
    ≤ 0.05 %
  • Zinc (Zn):
    ≤ 0.03 %

Specification Test Results

About this item

 :
 :237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:56 °C
Density:1,92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-589-4
UN:3077
ADR:9,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652