Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:13600-98-1
Formula:Na₃Co(NO₂)₆
MDL Number:MFCD00003512
Merck Index:13,08677
Melting Pt:∼220 °C
Storage Temperature:Ambient
EINECS:237-077-7
UN:1479
ADR:5.1,II
:403.94 g/mol
:GHS03
:GHS08
:GHS07
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≤0.02%
- For R&D use only. Not on TSCA Inventory.:Passes Test
- Insoluble Matter:≤0.02 %
- Suitability for Potassium Determination:Passes Test