Orders LinkContactUsLinkComponent
VWR®, 屈折計、デジタルハンドヘルド
VWR®, 屈折計、デジタルハンドヘルド
VWR®, 屈折計、デジタルハンドヘルド
VWR®, 屈折計、デジタルハンドヘルド
Dual-scale handheld refractometers designed to provide ranges of Brix (sugar concentration) and refractive index measurements.
  • Optical glass prism and stainless steel dish to stabilise sample temperature
  • Simple zero calibration with distilled water
  • Automatic temperature compensation (ATC) to correct readings of water and sucrose solutions to 20 °C
 : Supplied with two batteries and a battery indicator, multilingual guide and protective case.
  • Catalog No:
  • 635-0723
  • 635-0722
  • Accuracy:
  • ±0,0003 nD; ±0,2 °Bx
  • ±0,0003 nD; ±0,2 °Bx
  • Measurement time:
  • 2 s
  • 2 s
  • Resolution:
  • 0,1 °Bx; 0,0001 nD
  • 0,1 °Bx; 0,0001 nD
  • W×D×H:
  • 55×120×35 mm
  • 55×120×35 mm
  • Weight:
  • 85 g
  • 85 g
Product Family Options
  • 説明
    Range
    Refractive index
  • Digital handheld refractometer
    0 − 95 °Bx
    1,33 − 1,54 nD
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    635-0723
    Accuracy
    ±0,0003 nD; ±0,2 °Bx
    Measurement time
    2 s
    Resolution
    0,1 °Bx; 0,0001 nD
    W×D×H
    55×120×35 mm
    Weight
    85 g
  • Digital handheld refractometer
    0 − 54 °Bx
    1,33 − 1,42 nD
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    635-0722
    Accuracy
    ±0,0003 nD; ±0,2 °Bx
    Measurement time
    2 s
    Resolution
    0,1 °Bx; 0,0001 nD
    W×D×H
    55×120×35 mm
    Weight
    85 g