Orders LinkContactUsLinkComponent
解剖用鉗子、VWR®(ブイアール
解剖用鉗子、VWR®(ブイアール
Supplier:   VWR Collection
解剖用鉗子、VWR®(ブイアール
Supplier:  VWR Collection
Certificates
解剖用鉗子、VWR®(ブイアール
解剖用鉗子、VWR®(ブイアール
2 Options Available Below
Product Details & Documents
Dissecting forceps with extra fine tips. Made from stainless steel.
Product Family Options
Product Information
  • Type
    長さ
    Availability
    Price
  • Straight, very fine tips (tower tip)
    108 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $26.91
    $26.91
    1 * 1 items
       $26.91
  • Curved, very fine tips
    108 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $28.85
    $28.85
    1 * 1 items
       $28.85