Orders LinkContactUsLinkComponent
漏斗、粉体、VWR®製
漏斗、粉体、VWR®製
Supplier:   VWR Collection
漏斗、粉体、VWR®製
Supplier:  VWR Collection
Certificates
漏斗、粉体、VWR®製
漏斗、粉体、VWR®製
3 Options Available Below
Product Details & Documents
PP, translucent.
  • Parallel stem reduces powder bridging
  • External ribs prevents airlock
  • Resistant to most chemicals except strong oxidizing agents
  • Autoclavable
Used for transferring powders. Used from 0 to 135 °C.
Product Family Options
Product Information
  • Funnel O-Ø
    長さ
    Stem O-Ø
    Availability
    Price
  • 79 mm
    80,0 mm
    16,0 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $116.92
    $116.92
    1 * 12 items
       $116.92
  • 100 mm
    106,0 mm
    21,0 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $74.85
    $74.85
    1 * 6 items
       $74.85
  • 147 mm
    136,0 mm
    27,0 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items - $60.93
    $60.93
    1 * 4 items
       $60.93