Orders LinkContactUsLinkComponent
VWR®, 顕微鏡カバー
VWR®, 顕微鏡カバー
Supplier:   VWR Collection
VWR®, 顕微鏡カバー
Supplier:  VWR Collection
Certificates
VWR®, 顕微鏡カバー
VWR®, 顕微鏡カバー
6 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • 説明
    Availability
    Price
  • Dust cover, XL, for BI and TR 500 PH, 500×530 mm
    1 * 1 items
  • Dust cover, XXL, for BI and TR 500 PH, 600×530 mm
    1 * 1 items
  • Dust cover, large, L×H: 450×480 mm
    1 * 1 items
  • Dust cover, medium, L×H: 600×500 mm
    1 * 1 items
  • TNT dust cover, large, L×H: 700×550 mm (for models SZB360 OH, SZT360 OH and IT400 series)
    1 * 1 items
  • Dust cover, XL, L×H: 820×550 mm