ガウン 最大
About this item
These gowns are the cleanest and most durable within the entire VWR® Protection line of apparel. Gowns provide barrier protection against fluids, biologicals, particulates, and light chemical splash.
- Full protection from fluids and light chemical splash
- Manufactured from fluid-impervious material
- Lowest level of particle shedding
- Clean, low-linting fabric
- Elastic cuffs and ties at the neck and waist ensure secure closure
- Serged seams
- Bacterial filtration efficiency: 99,9%
Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Gowns are rigorously tested and manufactured in an ISO certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
5 Options Available Below
Product Details & Documents
These gowns are the cleanest and most durable within the entire VWR® Protection line of apparel. Gowns provide barrier protection against fluids, biologicals, particulates, and light chemical splash.
- Full protection from fluids and light chemical splash
- Manufactured from fluid-impervious material
- Lowest level of particle shedding
- Clean, low-linting fabric
- Elastic cuffs and ties at the neck and waist ensure secure closure
- Serged seams
- Bacterial filtration efficiency: 99,9%
Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Gowns are rigorously tested and manufactured in an ISO certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Specifications for Product Family
Specifications
- Catalog No:
- VWRU414004-681
- VWRU414004-470
- VWRU414004-471
- VWRU414004-682
- VWRU414004-469
- Closure type:
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Seam type:
- Serged
- Serged
- Serged
- Serged
- Serged
- Garment application:
- Pharmaceutical|Laboratory|Industrial|Food
- Pharmaceutical|Laboratory|Industrial|Food
- Pharmaceutical|Laboratory|Industrial|Food
- Pharmaceutical|Laboratory|Industrial|Food
- Pharmaceutical|Laboratory|Industrial|Food
- Bacterial filtration efficiency:
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
Specifications
Product Family Options
Product Information
- Colour衣服のサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.VWRU414004-681Closure typeBack fasteningGenderUnisexSeam typeSergedGarment applicationPharmaceutical|Laboratory|Industrial|FoodBacterial filtration efficiency99,9% - Specifications:
More
Specifications
Cat. No.VWRU414004-470Closure typeBack fasteningGenderUnisexSeam typeSergedGarment applicationPharmaceutical|Laboratory|Industrial|FoodBacterial filtration efficiency99,9% - Specifications:
More
Specifications
Cat. No.VWRU414004-471Closure typeBack fasteningGenderUnisexSeam typeSergedGarment applicationPharmaceutical|Laboratory|Industrial|FoodBacterial filtration efficiency99,9% - Specifications:
More
Specifications
Cat. No.VWRU414004-682Closure typeBack fasteningGenderUnisexSeam typeSergedGarment applicationPharmaceutical|Laboratory|Industrial|FoodBacterial filtration efficiency99,9% - Specifications:
More
Specifications
Cat. No.VWRU414004-469Closure typeBack fasteningGenderUnisexSeam typeSergedGarment applicationPharmaceutical|Laboratory|Industrial|FoodBacterial filtration efficiency99,9%