マルチロッドコネクター、Talon®
13 mm
VWRU60079-053
Multiple rod connector holds lab-frame rods or extension clamps at 90° while allowing both horizontal and vertical adjustment of lattice after construction. Two adjacent rods fit into oval hole in connector that prevent turning when tightened.
- Versatility and maximum adjustment capability at your fingertips
- Create a variety of configurations via side and front holes
: Two adjacent rods has to be ordered separately.
- For rod Ø:13 mm