Orders LinkContactUsLinkComponent
デシケーターキャビネット、アクリル製、VWR®
デシケーターキャビネット、アクリル製、VWR®
デシケーターキャビネット、アクリル製、VWR®
デシケーターキャビネット、アクリル製、VWR®
These acrylic desiccator cabinets can be used in refrigerators or at room temperature for dry storage of moisture-sensitive materials.
  • Fully removable, tray-like shelves have perforated bottoms and slide in/out on pre-set rails
  • Door opens from right to left and is secured with a stainless steel latch
Acrylic desiccator cabinets are fabricated from either clear or bronze acrylic, with a variable number of trays depending on model. Clear acrylic for easy viewing or bronze for light-sensitive materials. Bronze acrylic features stainless steel twist locking latches and hinges.
Product Family Options
  • 説明
    Shelves
    W×D×H ext.
  • Desiccator cabinet, clear acrylic, 23,2 L
    3
    305×305×305 mm
    1 * 1 items
    1 * 1 items
  • Desiccator cabinet, bronze acrylic, 23,2 L
    3
    305×305×305 mm