VWR®TraceClean®, Water sampling bottles, hdpe, cylindrical
VWRU89094-072
- 容量:250 ml
- 説明:Not Processed (NPC) HDPE bottle with poly-foam lined cap
- Seq:01
- Pk:24
HDPE, translucent.
- Available with poly-foam lined caps
- For use in metals and multiple inorganic analyses.
Processed (PC) and Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904