ウォーターバスクランプ、VWR® Talon®
VWRU21573-129
- Spanning width:5 - 10 mm
- 説明:Water bath clamp, small, two pronged
- 長さ:89 mm
- Pk:1
Hold a variety of apparatus, including glass tubes and thermometers, on glass water bath walls of varying thickness up to 9 mm (¹¹/₃₂"). Knurled thumbscrew tightens jaws to hold object firmly.