クランプホルダー、Talon®(タロン
0 - 19 mm
Catalog # VWRU21572-658
Supplier: VWR Collection
Jump to:
Product Details & Documents
Building on the upgraded Q-clamp design, this all-position clamp holder allows adjustment at any angle in any plane.
- Holders are set at 90° to each other connected by a 90° connector for 360° rotation
Specifications
- Spanning width:0 - 19 mm