スイベルホルダー、Talon®, VWR®
Catalog # VWRU21572-603
Supplier: VWR Collection
Specifications
- Spanning width:19 mm
- Pk:1
Specifications
About this item
This Q-clamp post holder with center swivel capacity allows tilting of clamps at any angle in parallel planes. Made of nickel-plated zinc.
- Allows tilting at any angle
- Outside adjustment screw allows close proximity between objects being held