Orders LinkContactUsLinkComponent
クランプホルダー、Talon®(タロン
クランプホルダー、Talon®(タロン
Supplier:   VWR Collection
クランプホルダー、Talon®(タロン
Supplier:  VWR Collection
Certificates
クランプホルダー、Talon®(タロン
クランプホルダー、Talon®(タロン
2 Options Available Below
Product Details & Documents
Features a streamlined design for maximum contamination resistance and easy cleaning.
  • Use wherever clamping at 90° is required
  • Available in nickel-plated zinc or electropolished stainless steel finish
  • Strong and durable
Regular clamp holders are used to hold extension clamps to lab frames where 90° positioning is required.
Product Family Options
Product Information
  • 説明
    Spanning width
    Material
    Availability
    Price
  • Clamp holder, regular
    18 mm
    Nickel-plated zinc
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $39.55
    $39.55
    1 * 1 items
       $39.55
  • Clamp holder, regular
    18 mm
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $172.30
    $172.30
    1 * 1 items
       $172.30