VWR®, Crucible Tongs, Stainless steel, with Corrugated Tip
VWRU89259-892
- 長さ:508 mm
- Pk:1
- Type:Corrugated tips
Stainless steel. These crucible holders have tight-fitting corrugated tips for handling dishes in muffle furnaces.
Constructed from a stainless steel rod with a diameter of 9 mm. There is a joint near the middle for holding large-sized crucibles.