クリーンルーム用フード, 照射済み
White, One size, +
Catalog # VWRU414004-502
Supplier: VWR Collection
Jump to:
Product Details & Documents
This irradiated hood is manufactured from a specially formulated breathable microporous fabric that provides significantly fluid and barrier protection.
- Excellent water vapour transmission to optimise user comfort
- Hood is gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁴ to address the concern of bio-burden contamination
- Lowest level of particle shedding
- Durable yet soft, cloth-like fabric
Packaging: Packed individually in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Specifications
- 衣服のサイズ:One size
- Sterile:+
- Colour:White
- Cat. No.:VWRU414004-502
- Cleanroom class:100000/ISO 8
- Garment application:Cleanroom