Orders LinkContactUsLinkComponent
ヒュームフード・ラボフレームキット、Talon®(タロン
ヒュームフード・ラボフレームキット、Talon®(タロン
Supplier:   VWR Collection
ヒュームフード・ラボフレームキット、Talon®(タロン
Supplier:  VWR Collection
Certificates
ヒュームフード・ラボフレームキット、Talon®(タロン
ヒュームフード・ラボフレームキット、Talon®(タロン
2 Options Available Below
Product Details & Documents
Rods are made of stainless steel. Connectors and lab frame feet are constructed of nicke plated zinc.
The 1,2 m kit includes four 965 mm horizontal rods, four 914 mm vertical rods, two 305 mm base rods, two 457 mm rear support rods, eight 51 mm rods, 30 lattice connectors, four rod end connectors and four lab frame feet. The 1,8 m kit includes four 1575 mm horizontal rods, six 914 mm vertical rods, two 305 mm base rods, two 457 mm rear support rods, eight 51 mm rods, 38 lattice connectors, four rod end connectors and four lab frame feet.
Product Family Options
Product Information
  • 説明
    Availability
    Price
  • For 1,2 m fume hoods
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $2,490.68
    $2,490.68
    1 * 1 items
       $2,490.68
  • For 1,8 m fume hoods
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $2,904.25
    $2,904.25
    1 * 1 items
       $2,904.25