スプレーボトル、 調整可能、VWR®
240 ml, Adjustable spray wash bottle
Catalog # VWRU23609-182
Supplier: VWR Collection
Jump to:
Product Details & Documents
HDPE with PP spray cap.
- Large capacity minimises refilling
- Chemical-resistant and virtually unbreakable
- Gas or chemically sterilised and dishwasher safe
- Spray pattern can be controlled by a contoured spray trigger with 'instant off'
These wash bottles with adjustable-spray stream keep the contents completely free of contaminants. Virtually closed system eliminates drips and handles hot, cold, aqueous and solvent-based solutions.
Specifications
- 容量:240 ml
- 説明:Adjustable spray wash bottle
- Seq:01