Orders LinkContactUsLinkComponent
分光光度計キュベット、VWR®
分光光度計キュベット、VWR®
分光光度計キュベット、VWR®
分光光度計キュベット、VWR®
Accurate and dependable results for spectrophotometric analyses in the visible and near infrared regions.
  • Matched sets within the accepted pathlength standard of ±0,01 mm
  • Cells exhibit a transmission tolerance within 0,5% at 365 nm
  • Lids and stoppers are made of PTFE
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the two optical windows.
Product Family Options
  • 説明
    Range
    高さ
    容量
    Path length
    Window material
  • Cells with flat lid, 12,5×12,5 mm
    334 – 2500 nm
    45 mm
    3,5 ml
    10 mm
    Optical glass
    1 * 2 items
    1 * 2 items - $225.09
    $225.09
    1 * 2 items
       $225.09
  • Cells with flat lid, 12,5×12,5 mm
    334 – 2500 nm
    45 mm
    3,5 ml
    10 mm
    Optical glass
    1 * 4 items
    1 * 4 items - $455.60
    $455.60
    1 * 4 items
       $455.60