VWR®, Glove Box Holder
611-3119
- Colour:White
- 説明:Glove boxes holder
- W×D×H:254×156×97 mm
- Pk:3
Glove box holder in durable resin to resist harsh cleaning solutions.
- Wall-mountable design for easy use
- Modular design for horizontal stacking of multiple boxes
- Universal fit for many of the major brands
Suitable for use in labs, cleanrooms, hospitals and microbiological working areas.
: Mounting screws included.