フックコネクター, Talon®
Hook connector, 13 mm, Stainless steel
Catalog # VWRU12985-062
Supplier: VWR Collection
Jump to:
Product Details & Documents
This hook connector allows the one-handed assembly of two components with one adjustment screw.
- Simple, versatile, and easy-to-use
- Maximum grip size: 13 mm (¹/₂")
Specifications
- Spanning width:13 mm
- 説明:Hook connector
- Material:Stainless steel
- Seq:01